Learn More
Medchemexpress LLC Manidipine | 89226-50-6 | 99.9% | 10 MM 1 ML
SDP
Supplier:  Medchemexpress LLC HYB041910MM/1ML
Encompass_Preferred
Manidipine is an orally active calcium channel antagonist. It regulates the expression of cytokines (IL-1β, IL-6) and has a hypotensive effect. It can be used in the study of cardiovascular diseases (such as hypertension, myocardial ischemia-reperfusion injury, ventricular hypertrophy), kidney diseases (such as glomerular diseases), and epilepsy.
  • Orally active calcium channel antagonist
  • Regulates expression of cytokines (IL-1β, IL-6)
  • Has a hypotensive effect
  • Used in studies of cardiovascular diseases, kidney diseases, and epilepsy
  • Inhibits transcription of mRNA for IL-1β and GM-CSF
  • Reduces systolic blood pressure in animal models
Catalog No. 50-004-09244