Learn More
Medchemexpress LLC HY-101603 1mg Medchemexpress, Manitimus CAS:202057-76-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1016031MG
Encompass_Preferred
Medchemexpress, HY-101603 1mg Manitimus CAS:202057-76-9 Manitimus is an inhibitor of dehydroorotate dehydrogenase, and a potent immunosuppressive drug. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7860