Learn More
General Laboratory Products Mannitol Salt Agar, 500 mL, 12/cs
Supplier:  General Laboratory Products 500EZ7116500
Encompass_Preferred
Mannitol Salt Agar - 500ml, Easy-pour narrow mouth glass bottle, 12 per case. Base for the isolation of Staphylococci.
Catalog No. NC0503079