Learn More
Grainger Manual AirControl 3-Way 2-Position 10-32
Supplier:  Grainger 14F274
Encompass_Preferred
The Pneumadyne Inc. Manual Air Control Valve Three-Way is a manual air control valve with three ports that allows you to control the direction of air flow. It has a brass body construction with a chrome-plated finish and features a knurled knob for easy hand operation. This three-way valve enables you to route air flow between two output ports or block the air flow completely. It has a maximum pressure rating of 250 psi and temperature range of -20 to 200 degrees Fahrenheit. The valve has 1/4" NPT thread ports and includes a mounting bracket for installation. This air valve provides precise manual air flow control for pneumatic applications.
Catalog No. 50-275-0787