Learn More
Chemglass Life Sciences Crimper, Hand Operated, 20mm
SDP
Supplier:  Chemglass Life Sciences CG493020
Encompass_Preferred
Crimper, Hand Operated, 20mm. Tool for crimping Aluminum seals on Microwave vials. The crimper has an adjustment feature which will allow it to be used with a variety of septa, discs, stoppers or lined caps. The user may adjust the stroke or movement for each application as needed. For Microwave Vials and accessories, see CG-4920.
Catalog No. 50-872-709