Learn More
World Precision Instrument Manual Microsyringe Pump-DMP
SDP
Supplier:  World Precision Instrument DMP
Encompass
The manual microsyringe pump is a convenient tool for precise manual injection of fluid using glass pipettes or similar injection devices. The design allows visual feedback of flow at the pipette tip. They can also be used as a manual micro syringe pump for perfusion or withdrawal of liquids. The resolution of the injection volume can be continuously varied from 10 nanoliters to the microliter range, depending on the syringe used.
Catalog No. 50-252-1195