Learn More
AdipoGen Manumycin B
SDP
Supplier:  AdipoGen BVT0264M001
Encompass_Preferred
Chemical. CAS 139023-58-8. Formula C28H34N2O7. MW 510.6. Isolated from Streptomyces parvulus. Antibiotic. Antibacterial. Active against Gram-positive bacteria. Rasfarnesyltransferase inhibitor. Apoptosis Caspase-1 inhibitor. AChE inhibitor.
Catalog No. 50-256-2025