Learn More
Hach Company ManVer 2 Hardness Indicator Powder, 113g
Supplier:  Hach Company 28014
Encompass
Indicator for hardness determination by EDTA titration. Bulk Powder. APHA Hardness Method. 113g jar
Catalog No. 50-199-4232