Learn More
Medchemexpress LLC Manzamine A 1mg | 104196-68-1 | 548.76 | C36H44N4O | 1 MG

Supplier: Medchemexpress LLC HY1170251MG
Manzamine A is a marine-derived β-carboline alkaloid provided as a research reagent. It is reported to inhibit GSK-3β and CDK-5 and has been investigated for antimalarial and anticancer activity. Offered as the free base (CAS 104196-68-1), this material is intended for laboratory research use only.
- Purity 98.2% (by vendor specification).
- Chemical formula C36H44N4O; molecular weight ≈ 548.76 g/mol.
- Available as a 1 mg research pack.
- Free base form; hydrochloride salt available separately.
- For research use only; not for human or clinical use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.