Learn More
Discovery Life Sciences MAO A SUPERSOME
SDP
Supplier:  Discovery Life Sciences 456283
Encompass
Human Monoamine Oxidase A (MAO-A) 0.5mL Catalyzed by MAO-A which is expressed from human MAO-A cDNA using a baculovirus expression system. Baculovirus infected insect cells (BTI-TN-5B1-4) were used to prepare these membranes
Catalog No. NC9906032