Learn More
Medchemexpress LLC Mao-B-IN-30 | 82973-15-7 | 98.5% | 344.16 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY161240100MG
Encompass_Preferred
MAO-B-IN-30 (compound IS7) is a potent, selective, and blood-brain barrier-crossing MAO-B inhibitor with IC50 values of 19.176 μM for MAO-A and 0.082 μM for MAO-B. It exhibits antiproliferative activity and is non-cytotoxic. MAO-B-IN-30 reduces levels of TNF-alpha, IL-6, and NF-kB, and shows potential for Parkinson's disease research.
  • Potent, selective, and blood-brain barrier-crossing MAO-B inhibitor.
  • Shows antiproliferative activity and is non-cytotoxic.
  • Reduces TNF-alpha, IL-6, and NF-kB levels.
  • Potential for Parkinson's disease research.
  • For research use only.
Catalog No. 50-003-92638