Learn More
Medchemexpress LLC MAO-B-IN-30 | 82973-15-7 | 98.5% | 344.16 | 5MG
SDP
Supplier:  Medchemexpress LLC HY1612405MG
Encompass_Preferred
MAO-B-IN-30 (compound IS7) is a potent, selective, and blood-brain barrier-crossing MAO-B inhibitor. It exhibits IC50 values of 19.176 μM for MAO-A and 0.082 μM for MAO-B, indicating its high selectivity for MAO-B. This compound also demonstrates antiproliferative activity while being non-cytotoxic. Furthermore, MAO-B-IN-30 has been shown to reduce levels of TNF-alpha, IL-6, and NF-kB. These characteristics suggest its potential for research into Parkinson's disease.
  • It is a potent and selective MAO-B inhibitor.
  • It can cross the blood-brain barrier, making it suitable for neurological research.
  • It shows antiproliferative activity.
  • It is non-cytotoxic, indicating a favorable safety profile in certain contexts.
  • It reduces TNF-alpha, IL-6, and NF-kB levels, suggesting anti-inflammatory properties.
  • It has potential for Parkinson's disease research.
Catalog No. 50-003-92920