Learn More
TARGETMOL CHEMICALS INC MAO-B-IN-9 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T606715MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. MAO-B-IN-9 is a monoamine oxidase B MAO-B inhibitor (IC50 0.18 uM) that crosses the blood-brain barrier with potency selectivity and time-dependence. MAO-B-IN-9 has neuroprotective effects against A(beta)1-42-induced neuronal cell death which may be related to the anti-aggregation effect of A(beta)1-42. purity: 99%
Catalog No. 50-257-9595