Learn More
Medchemexpress LLC Mao-in-m30 dihydrochloride | 64821-19-8 | 98.56% | 50 MG
SDP
Supplier:  Medchemexpress LLC HY13103650MG
Encompass_Preferred
MAO-IN-M30 dihydrochloride is an orally active, brain-permeable, and brain-selective irreversible inhibitor of MAO-A (IC50=37 nM) and MAO-B (IC50=57 nM). It acts as a potent iron chelator and radical scavenger, demonstrating neuroprotective effects against Dexamethasone-induced brain cell apoptosis. Additionally, it exhibits neurorestorative activity in post-MPTP and lactacystin models of Parkinson's disease. This compound is also a click chemistry reagent.
  • Orally active and brain-permeable irreversible inhibitor of MAO-A and MAO-B
  • Potent iron chelator and radical scavenger
  • Demonstrates neuroprotective effects against dexamethasone-induced brain cell apoptosis
  • Exhibits neurorestorative activity in post-MPTP and lactacystin models of Parkinson's disease
  • Click chemistry reagent containing an alkyne group
Catalog No. 50-003-40582