Learn More
Medchemexpress LLC Mao-in-m30 dihydrochloride | 64821-19-8 | 98.6% | 100 MG
SDP
Supplier:  Medchemexpress LLC HY131036100MG
Encompass_Preferred
MAO-IN-M30 dihydrochloride is an orally active, brain-permeable, and brain-selective irreversible MAO-A and MAO-B inhibitor. This potent iron chelator and radical scavenger exhibits a neuroprotective effect against dexamethasone-induced brain cell apoptosis. It also demonstrates neurorestorative activity in models of Parkinson's disease and is a click chemistry reagent.
  • Orally active, brain-permeable, and brain-selective
  • Irreversible MAO-A (IC50=37 nM) and MAO-B (IC50=57 nM) inhibitor
  • Potent iron chelator and radical scavenger
  • Exhibits neuroprotective effect against dexamethasone-induced brain cell apoptosis
  • Demonstrates neurorestorative activity in Parkinson's disease models
  • Click chemistry reagent with an alkyne group, capable of CuAAc reactions
Catalog No. 50-003-39030