Learn More
Aviva Systems Biology Corp MAP2K2 Antibody - N-terminal region (ARP48941_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48941_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MAP2K2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human MAP2K2. Alternate names: MAPKK2, MEK2, MKK2, PRKMK2ApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rat
Catalog No. 50-014-59967