Learn More
ABclonal Technology MAP3K14 Rabbit pAb
SDP
Supplier:  ABclonal Technology A115851000UL
Encompass_Preferred
This gene encodes mitogen-activated protein kinase kinase kinase 14, which is a serine/threonine protein-kinase. This kinase binds to TRAF2 and stimulates NF-kappaB activity. It shares sequence similarity with several other MAPKK kinases. It participates in an NF-kappaB-inducing signalling cascade common to receptors of the tumour-necrosis/nerve-growth factor (TNF/NGF) family and to the interleukin-1 type-I receptor.
Catalog No. 50-001-14509