Learn More
Aviva Systems Biology Corp MAP4K2 Antibody - N-terminal region (ARP48632_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48632_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MAP4K2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human MAP4K2. Alternate names: BL44, GCK, RAB8IPApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59872