Learn More
Aviva Systems Biology Corp MAP6 Antibody - N-terminal region (ARP60316_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60316_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MAP6. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human MAP6. Alternate names: FLJ41346, KIAA1878, MTAP6, N-STOP, STOPApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-287-003