Learn More
Medchemexpress LLC Maprotiline (hydrochloride) | 10347-81-6 | 100.0% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB044410MM/1ML
Encompass_Preferred
Maprotiline hydrochloride is a highly selective noradrenergic reuptake inhibitor with strong antidepressant, antitumor, and neuropathic pain-relieving effects. It is orally active and induces cancer cell apoptosis by targeting the ERK signaling pathway and CRABP1.
  • Highly selective noradrenergic reuptake inhibitor
  • Exhibits antidepressant, antitumor, and neuropathic pain-relieving effects
  • Orally active
  • Induces cancer cell apoptosis by targeting the ERK signaling pathway and CRABP1
  • Inhibits growth and metastasis of liver cancer cells
  • Regulates cholesterol biosynthesis
  • Inhibits tumor growth in liver cancer mouse models without obvious toxicity
  • Attenuates neuropathic pain in mice
Catalog No. 50-004-05443