Learn More
Medchemexpress LLC Maprotiline hydrochloride | 10347-81-6 | 100.0% | 100 MG
Supplier: Medchemexpress LLC HYB0444100MG
Maprotiline hydrochloride is a highly selective noradrenergic reuptake inhibitor with potent antidepressant, antitumor, and neuropathic pain-relieving effects, demonstrating oral activity. It induces cancer cell apoptosis by targeting the ERK signaling pathway and CRABP1.
- Inhibits growth and metastasis of Huh7 and HepG2 cells
- Triggers cell apoptosis
- Affects the ERK signaling pathway
- Regulates cholesterol biosynthesis in HCC cells by targeting CRABP1
- Suppresses liver cancer tumor growth in mice
- Reduces neuropathic pain in mice