Learn More
Medchemexpress LLC Maprotiline hydrochloride | 10347-81-6 | ≥98% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYB0444R
Encompass_Preferred
Maprotiline hydrochloride is a highly selective noradrenergic reuptake inhibitor, demonstrating strong antidepressant, antitumor, and neuropathic pain-relieving effects through oral activity. This substance induces cancer cell apoptosis by targeting the ERK signaling pathway and CRABP1. It is a solid with a molecular formula of C20H24ClN and a molecular weight of 313.86, having a melting/freezing point of 230-232°C.
  • Highly selective noradrenergic reuptake inhibitor
  • Strong antidepressant effects
  • Antitumor properties
  • Relieves neuropathic pain
  • Induces cancer cell apoptosis
  • Targets ERK signaling pathway
  • Targets CRABP1
Catalog No. 50-004-50586