Learn More
Medchemexpress LLC Maprotiline (hydrochloride) | 10347-81-6 | 99.9% | 500 MG
SDP
Supplier:  Medchemexpress LLC HYB0444500MG
Encompass_Preferred
Maprotiline hydrochloride is a highly selective noradrenergic reuptake inhibitor with strong antidepressant, antitumor, and neuropathic pain-relieving effects, exhibiting oral activity. This compound induces cancer cell apoptosis by targeting the ERK signaling pathway and CRABP1.
  • Induces cell apoptosis and inhibits cell viability in a dose- and time-dependent manner.
  • Inhibits metastasis and restrains migration of liver cancer cells.
  • Affects the ERK pathway and inhibits SREBP2 phosphorylation.
  • Targets CRABP1 and regulates cholesterol biosynthesis.
  • Effectively reduces neuropathic pain in mice.
  • Inhibits liver cancer tumor growth in vivo.
Catalog No. 50-004-08448