Learn More
Selleck Chemical LLC Maraviroc 25mg 376348-65-1 UK-427857
SDP
Supplier:  Selleck Chemical LLC S200325MG
Encompass_Preferred
Maraviroc (UK-427857) is a CCR5 antagonist for MIP-1?, MIP-1? and RANTES with IC50 of 3.3 nM, 7.2 nM and 5.2 nM in cell-free assays, respectively. Maraviroc is used in the treatment of HIV infection. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-705-4