Learn More
Cayman Chemical MarcfortIn A 5mg
SDP
Supplier:  Cayman Chemical 234865 MG
Encompass_Preferred
An anthelmintic; has nematocidal activity against H. contortus (LD99 = 0.06 μg/ml) and inhibits motility of adult worms (EC50 = 2 μM); eliminates H. contortus, T. colubriformis, and O. ostertagi from experimentally infected jirds (ED95s = 0.33, 0.11, and 2.5 mg/animal, respectively); dose-dependently inhibits nicotine-induced calcium mobilization in SH-SY5Y and TE-671 cells expressing α3 subunit-containing and muscle-type nAChRs, respectively
Catalog No. 50-299-4601