Learn More
Legend Inc Marcy Pulp Density Scale (container sold separately)
SDP
Supplier:  Legend Inc 18380
Encompass
Greater Sensitivity: Circular dial is equivalent to a 47" beam balance for greater sensitivity.Solid Construction: Entire unit is supported by a top ring. Scale is accurately set at the factory.Accuracy: Cylindrical containers with slotted overflow, permit accurate filling to exactly 1000 cc.Ease Of Operation: Samples do not have to be weighed separately; refer to tables or make calculations.Compatibility: Interchangeable dials make weighing applicable for any range of solids of light or heavy density. APPLICATIONS:Use Marcy Scales to obtain the following direct readings: Weight of sample in grams or kilograms. Specific gravity of liquid or pulp. Results are in % solids by weight. Percent solids contained in the pulp of any given specific gravity.
Catalog No. NC9726402