Learn More
Medchemexpress LLC Mardepodect | 898562-94-2 | 98.8% | C25H20N4O | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY50098
Encompass_Preferred
Mardepodect (PF-2545920) is a potent, selective orally active, and brain-penetrant PDE10A inhibitor with an IC50 of 0.37 nM, demonstrating over 1000-fold selectivity over other PDEs. It has shown activity in preclinical models for positive, cognitive, and negative symptoms of schizophrenia and increases striatal cGMP in mice in a dose-dependent manner. This product is for research use only.
  • Potent and selective PDE10A inhibitor
  • Orally active and brain-penetrant
  • IC50 of 0.37 nM
  • Exhibits >1000-fold selectivity over other PDEs
  • Active in conditioned avoidance response (CAR) assay with an ED50 of 1 mg/kg
  • Causes a dose-dependent increase in striatal cGMP in mice
Catalog No. 50-003-58548