Learn More
Medchemexpress LLC Mardepodect hydrochloride | 2070014-78-5 | 99.9% | C25H21ClN4O | 1 ML
SDP
Supplier:  Medchemexpress LLC HY50098A10MM/1ML
Encompass_Preferred
Mardepodect hydrochloride is a potent, orally active, and selective PDE10A inhibitor with an IC50 of 0.37 nM. It demonstrates over 1000-fold selectivity compared to other PDEs and can cross the blood-brain barrier. This product is for research use only and is not sold to patients.
  • Potent, orally active, and selective PDE10A inhibitor
  • >1000-fold selectivity over other PDEs
  • Capable of crossing the blood-brain barrier
  • Active in conditioned avoidance response assays (ED50 of 1 mg/kg)
  • Administration to mice increases striatal cGMP dose-dependently
Catalog No. 50-003-57734