Learn More
Cayman Chemical MaresIn 1-d5 100ug
SDP
Supplier:  Cayman Chemical 21823100 UG
Encompass_Preferred
An internal standard for the quantification of maresin 1 by GC- or LC-MS
Catalog No. 50-298-6087