Learn More
Medchemexpress LLC HY-10985 100ug Medchemexpress, Marizomib CAS:437742-34-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10985100UG
Encompass_Preferred
Medchemexpress, HY-10985 100ug Marizomib CAS:437742-34-2 Marizomib (Salinosporamide A) is a second-generation, irreversible, brain-penetrant, pan-proteasome inhibitor. Marizomib inhibits the CT-L (β5), CT-T-laspase-like (C-L, β1) and trypsin-like (T-L, β2) activities of the 20S proteasome (IC50=3.5, 28, and 430 nM, respectively)[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8723