Learn More
Microsolv Technology Corporation Marker, permanent. Extra-Fine tip, 0.25mm. Silver Ink for plasticware. 1 EA.
Supplier:  Microsolv Technology Corporation 95296MARKFS
Encompass_Preferred
Mark in bold, opaque silver color on projects and more with this marker. The Extra Fine tip features a 0.7 mm pin-type plastic tip andValve-action with rich, smooth-flowing paint. Archival quality, waterproof, and fade resistant on most surfaces including, plastic, wood, porcelain, paper and metal. Permanent adhesion on most surfaces- except for glass and ceramics. Low-odor and xylene free. Meets ASTM and ACMI non-toxicity standards.
Catalog No. 50-227-0004