Learn More
MSC Marlin Steel Mesh Basket, Stainless Steel, With Lid, #6 Mesh, Electropolished
Supplier:  MSC 70699756
Encompass
This mesh basket is designed for efficient parts washing, material handling, and ultrasonic cleaning applications. Constructed from durable stainless steel, it features an electropolished finish for a smooth, nonstick surface, and a secure lid and latch for enclosed operation. Its #6 mesh design ensures optimal liquid and air flow.
  • Includes a lid and latch
  • Electropolished for a smooth, shiny finish
  • Features #6 mesh openings for effective flow
  • Suitable for parts washing and ultrasonic cleaning
  • Ideal for material handling applications
  • Made from durable stainless steel
  • Resistant to corrosion
Catalog No. 50-005-27710