Learn More
AdipoGen Martinomycin (1 mg)
SDP
Supplier:  AdipoGen AGCN20159M001
Encompass_Preferred
Chemical. CAS: 160791-16-2. Formula: C49H84O17. MW: 945.2. Isolated from Actinoallamurus yoronensis. Antibiotic. Active against Gram-positive bacteria. Active against Spodoptera eridania.
Catalog No. 50-205-3546