Learn More
Thermo Scientific™ MAS™ Liquimmune™ Immunoassay Controls
GSA_VA
Get relief from time consuming and costly QC standard lot changes for up to 5 years with ready-to-use, liquid assayed Thermo Scientific™ MAS™ Liquimmune™ Immunoassay Controls.
Products 6
Catalog Number Mfr. No. Description dimLevel Price Quantity & Availability  
Catalog Number Mfr. No. Description dimLevel Price Quantity & Availability  
23-139-403
GSA_VA
View Documents
Thermo Scientific™
10013876
N/A N/A
This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance.
23-139-404
GSA_VA
View Documents
Thermo Scientific™
10013877
N/A N/A
This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance.
23-139-405
GSA_VA
View Documents
Thermo Scientific™
10013878
N/A N/A
This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance.
LIG101
GSA_VA
Accessories View Documents
Thermo Scientific™
LIG101
N/A N/A
This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance.
LIG303
GSA_VA
Accessories View Documents
Thermo Scientific™
LIG303
N/A N/A
This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance.
LIG202
GSA_VA
Accessories View Documents
Thermo Scientific™
LIG202
N/A N/A
This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance.
Description
Description
Get relief from time-consuming and costly QC standard lot changes for up to 5 years with ready-to-use, liquid assayed Thermo Scientific™ MAS™ Liquimmune™ Immunoassay Controls. With a new 5-year shelf life, these multi-analyte controls simplify QC while monitoring immunoassay test procedures on most automated instruments. These controls are also available in a convenient Vista™ vial format. Extended claim may not apply for all countries.
  • Features three significant levels
  • Human serum matrix
  • Assayed values available for most automated instruments
  • Extended 5-year shelf life beginning with lot LIA2004 in Fall 2015
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.