Learn More
Thermo Scientific™ MAS™ Liquimmune™ Immunoassay Controls
GSA_VA
Get relief from time consuming and costly QC standard lot changes for up to 5 years with ready-to-use, liquid assayed Thermo Scientific™ MAS™ Liquimmune™ Immunoassay Controls.
Supplier:  Thermo Scientific™ LIG202
 View more versions of this product
Catalog No. LIG202
This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance.
This item is not returnable. View return policy
Description
Description
Get relief from time-consuming and costly QC standard lot changes for up to 5 years with ready-to-use, liquid assayed Thermo Scientific™ MAS™ Liquimmune™ Immunoassay Controls. With a new 5-year shelf life, these multi-analyte controls simplify QC while monitoring immunoassay test procedures on most automated instruments. These controls are also available in a convenient Vista™ vial format. Extended claim may not apply for all countries.
  • Features three significant levels
  • Human serum matrix
  • Assayed values available for most automated instruments
TRUSTED_SUSTAINABILITY
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Accessories