Learn More
Thermo Scientific™ MAS™ T-Marker™ Controls
Monitor the performance of various cancer antigen tests on automated instruments with liquid assayed, multi-analyte Thermo Scientific™ MAS™ T-Marker Controls.
| Catalog Number | Mfr. No. | dimLevel | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | dimLevel | Price | Quantity & Availability | |||||
|
TU1101
|
Thermo Scientific™
TUM101 |
N/A | N/A | ||||||
| This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance. | |||||||||
|
TU2202
|
Thermo Scientific™
TUM202 |
N/A | N/A | ||||||
| This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance. | |||||||||
|
TU3303
|
Thermo Scientific™
TUM303 |
N/A | N/A | ||||||
| This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance. | |||||||||
Description
Monitor the performance of various cancer antigen tests on automated instruments with liquid assayed, multi-analyte Thermo Scientific™ MAS™ T-Marker Controls. These controls contain more 24 analytes, including a complete offering of cancer-associated antigens.
- Liquid, ready to use; no additional preparation required
- Human serum matrix
- Includes CA 15-3, CA 19-9, CA 72-4, CA-125, CA 27-29
- Assayed values available for most major analyzers.