Learn More
Medchemexpress LLC Masitinib mesylate | 1048007-93-7 | MFCD12546334 | 99.9% | 594.75 g/mol | C29H34N6O4S2 | 50 MG

Supplier: Medchemexpress LLC HY10209A50MG
Masitinib mesylate is the methanesulfonate (mesylate) salt of masitinib, a selective, orally bioavailable tyrosine kinase inhibitor active against c-Kit, PDGFR, and Src family kinases. It is supplied as an analytical/research standard for in vitro and biochemical assays and is commonly used as a reference compound in kinase profiling and inhibitor studies.
- High purity suitable for analytical use.
- Molecular weight 594.75 g/mol.
- Chemical formula C29H34N6O4S2.
- Supplied as a 50 mg package.
- Intended for research and analytical applications.
- Useful as a reference standard in kinase assays and profiling.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.