Learn More
Grainger Masking Tape 1/2 W 1/2 L PK1000
Supplier:  Grainger 15C584
Encompass_Preferred
The TapeCase Film Tape Square is a 3-inch square of 2 mil polypropylene film tape. It has an acrylic adhesive and is transparent and glossy. The tape is designed for general purpose holding, patching, sealing, splicing, and masking applications. It bonds well to paper, plastic, glass, and metal. The operating temperature range is -20F to 200F. The tape comes in a handy dispenser box for easy dispensing and storage. Overall, this is a versatile, general use film tape that can be useful for a variety of household and industrial tasks.
Catalog No. 50-274-9806