Learn More
Grainger Masking Tape 2 W 2 L Transparent PK250
Supplier:  Grainger 15C587
Encompass_Preferred
The 3M Multi-Use Painters Masking Tape is a general use masking tape that can be used for holding, bundling, sealing, splicing, and masking. It has a medium adhesion level and is designed for 14 day use when applied to most surfaces. This 1.88 inch by 60 yard roll tape has a crepe paper backing and rubber adhesive. It can be used for both indoor and outdoor applications. The temperature use range is from 50 to 200 degrees Fahrenheit. This tape removes cleanly with no adhesive residue up to 14 days after application.
Catalog No. 50-274-9623