Learn More
Essendant MASKN95DISPOSABLE10WH
Supplier:  Essendant HWLDC300N95
Encompass_Preferred
NIOSH-Approved N95 use-once respirator provides excellent protection from dust mists and fumes in non-oil environments Contoured shape and molded nose bridge ensure wearing comfort Strong rigid outer shell prevents the mask from collapsing in high-humidity environments Dual-welded head strap Respirator Type- Half-Face Mask Rating- N95 Airborne Contaminant Resistance- Solid and Liquid Particulates Excluding Oils Material(s)- Electrostatic Non-Woven Polypropylene Polyester Fibers
Catalog No. 50-000-31574