Learn More
Essendant MASKRESPIRATORN95WE
Supplier:  Essendant MMM8210
Encompass_Preferred
A disposable particulate respirator designed to help provide reliable respiratory protection of at least 95 percent filtration efficiency against certain non-oil based particles Designed for particles such as those from grinding sanding sweeping sawing bagging or other dusty operations This respirator can also help reduce inhalation exposures to certain airborne biological particles (examples- mold Bacillus anthracis Mycobacterium tuberculosis) but cannot eliminate the risk of contracting infection illness or disease The respirator incorporates 3M s proprietary technology with advanced electrostatically charged microfiber filter media designed for ease of breathing Compatible with a variety of protective eyewear and hearing protection Breathing hazardous particles can pose a risk to your health Respirator Type- Half-Face Mask Rating- N95 Airborne Contaminant Resistance- Non-Oil Based Particulates Material(s)- Advanced Electrostatic Media
Catalog No. 50-000-31168