Learn More
Apexbio Technology LLC Maslinic Acid 4373-41-5 25mg
SDP
Supplier:  Apexbio Technology LLC C483225
Encompass_Preferred
Maslinic acid (CAS 4373-41-5) is a small-molecule inhibitor targeting inflammatory mediators such as nitric oxide (NO) hydrogen peroxide (H O ) interleukin-6 (IL-6) and tumor necrosis factor-alpha (TNF- ) It is designed to suppress production of these pro-inflammatory factors thereby modulating inflammation-associated signaling pathways Maslinic acid exerts its biological activity primarily by regulating apoptosis-related proteins promoting cytochrome-c release and activating caspase-9 and caspase-3 In cell-based studies using the HT29 colon cancer cell line maslinic acid demonstrates inhibitory effects on cell viability with an IC of 28 8 0 9 g/mL Based on these pharmacological properties maslinic acid holds research potential in oncology antiviral and metabolic disorder applications
Catalog No. 50-000-91465