Learn More
TARGETMOL CHEMICALS INC MASM7 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T500935MG
Encompass_Preferred
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. MASM7 also known as MASM7, is a compound used as a molecular structural unit. It is a mitochondrial fusion protein activator that enables mitochondrial fusion via mitochondrial fusion proteins. Purity 99.56%
Catalog No. 50-287-6706