Learn More
Medchemexpress LLC Masm7 | 920868-45-7 | 99.9% | 453.58 g/mol | C22H23N5O2S2 | 250 MG
SDP
Supplier:  Medchemexpress LLC HYW187305250MG
Encompass_Preferred
MASM7 is a small-molecule mitofusin activator that promotes mitochondrial fusion by directly activating mitofusins such as MFN2. It is supplied as a high-purity solid intended for research use in cellular and biochemical studies.
  • Promotes mitochondrial fusion via mitofusin activation.
  • Demonstrates cellular activity with an EC50 of about 75 nM in MEFs.
  • Binds the MFN2 HR2 domain with a reported Kd of approximately 1.1 μM.
  • High purity (>99.8%) suitable for research applications.
  • Molecular weight 453.58 g/mol and formula C22H23N5O2S2.
  • Supplied as a white to yellow solid; commonly available in 250 MG packs.
Catalog No. 50-002-81778