Learn More
Medchemexpress LLC Masoprocol (meso-nordihydroguaiaretic acid) | 27686-84-6 | 99.3% | 302.36 g/mol | C18H22O4 | 5 MG

Supplier: Medchemexpress LLC HY1095005MG
Masoprocol (meso-nordihydroguaiaretic acid) is a research-grade small molecule described as a lipoxygenase inhibitor with reported antihyperglycemic activity. Provided at milligram scale for in vitro and preclinical studies; not for human use.
- High purity: 99.3%.
- Molecular formula: C18H22O4.
- Molecular weight: 302.36 g/mol.
- Active as a lipoxygenase inhibitor and shows antihyperglycemic activity.
- Supplied in milligram-scale quantities suitable for laboratory research.
- Recommended storage: -20°C, protect from light; in solvent: -80°C up to 6 months, -20°C up to 1 month.
- Intended for research use only, not for human consumption.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.