Learn More
Medchemexpress LLC Massoia lactone | 54814-64-1 | 10 G
SDP
Supplier:  Medchemexpress LLC HYN743510G
Encompass_Preferred
Massoia lactone is a fragrant biosurfactant and a coconut and creamy fragrant compound. It is primarily isolated from Cryptocarya massoy and also produced by the fungus Aureobasidium pullulans.
  • High purity of 98.75%
  • Appears as a liquid
  • Color ranges from light yellow to light brown
  • Primarily isolated from Cryptocarya massoy
  • Also produced by the fungus Aureobasidium pullulans
  • Intended for research use only
Catalog No. 50-004-17430