Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex C-Flex ULTRA tubing, L/S 16, 25 ft.
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX0643416
Encompass
Masterflex C-Flex ULTRA tubing, L/S 16, 25 ft.Long life tubing for any Masterflex pump used in your pharma and bioprocess applicationsApplications: General-purpose lab uses requiring high performance and long life.Characteristics: Exceptional tensile and tear strength; good biocompatibility; low spallation, reduced residue and low gas permeability; and a smooth surface. Translucent.
Catalog No. NC0224455