Learn More
AVANTOR MASTERFLEX (VWR LLC) L/S Digital Drive, 100 RPM, 115/230 VAC
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX0752230
Encompass
Masterflex L/S Digital Drive, 100 rpm; 115/230 VACPrecision digital drive offers a maintenance-free high-accuracy motor and a graphical interface ¿ makes setup and operation easier than ever!Flow rate 0.001/0.006 to 566/3400 mL/min with L/S tubing¿flow rate depends on drive rpm, pump head, and tubing sizeGraphical LCD shows four operating modes continuous run, timed dispense, copy dispense, volume dispenseView pump performance continuallyDispense by Volume in mL (0.001 to 9,999 mL), L, or gal; Copy¿1 to 9,999 dispense cycles; or Time¿time interval
Catalog No. NC0611021