Learn More
Accuflow Systems Masterflex Pump Head, L/S Easy-Load II, Fixed Occlusion, 1 ea
Supplier:  Accuflow Systems MFLX7720060EA/DEL
Encompass_Preferred
Masterflex L/S Easy-Load II pump heads are designed for simple, easy-to-use peristaltic pump systems when combined with compatible Masterflex L/S drives. They accept various tubing sizes for a wide range of flow rates, making them suitable for laboratory fluid transfer applications.
  • Quick tubing changes enabled by over-center cam design and automatic tubing retention.
  • Self-priming and capable of dry operation without damage.
  • Compatible with most chemicals and feature no valves or seals.
  • Four rollers ensure smooth operation and accurate flow.
  • Max pump speed up to 600 RPM for efficient fluid handling.
  • Operating temperature range from 0°C to 40°C.
  • Constructed with polyphenylene sulfide and stainless steel for durability.
Catalog No. NC3948699