Learn More
HIGH PURITY NEW ENGLAND MASTERFLEX SNGL-USE QUATTROFLO
SDP
Supplier:  HIGH PURITY NEW ENGLAND MFLX7600001G/Q12448
Encompass
Masterflex® Single-Use Quattroflow™ Disposable Pump Chamber with EZ-Set, Polypropylene, 1/4" Tri-Clamp Gamma Irradiated
Catalog No. NC3934114